Learn More
Lascar Electronics Inc Temperature Data Logger with USB
SDP
Catalog No. NC9902721
Encompass_Preferred
This standalone USB temperature data logger can take more than 16,000 readings over a -35 to +80°C (-31 to +176°­­F) measurement range. Easily set up the logger and view downloaded data by plugging the unit into a PC’s USB port and using the free EasyLog software provided. Data can then be graphed, printed and exported to other applications for detailed analysis. Supplied with ½ AA battery and wall mount clip. Calibration certificate available to purchase separately.
Catalog No. NC9902721 Supplier Lascar Electronics Inc Supplier No. ELUSB1
This standalone USB temperature data logger can take more than 16,000 readings over a -35 to +80°C (-31 to +176°­­F) measurement range. Easily set up the logger and view downloaded data by plugging the unit into a PC’s USB port and using the free EasyLog software provided. Data can then be graphed, printed and exported to other applications for detailed analysis. Supplied with ½ AA battery and wall mount clip. Calibration certificate available to purchase separately.