Learn More
Lascar Electronics Inc Temperature Data Logger with USB and Display
SDP
Catalog No. NC0199480
Encompass_Preferred
This standalone data logger measures more than 16,000 readings over a -35 to +80°C (-31 to +176°¬-F) range. At the touch of a button, use the on-board display to cycle between the current, minimum and maximum temperature logs seen during the session.Easily set up the logger and view downloaded data by plugging the unit into a PC’s USB port and using the free EasyLog software provided. Data can then be graphed, printed and exported to other applications for detailed analysis. Supplied with ½ AA battery and wall mount clip.
Catalog No. NC0199480 Supplier Lascar Electronics Inc Supplier No. ELUSB1LCD
This standalone data logger measures more than 16,000 readings over a -35 to +80°C (-31 to +176°¬-F) range. At the touch of a button, use the on-board display to cycle between the current, minimum and maximum temperature logs seen during the session.Easily set up the logger and view downloaded data by plugging the unit into a PC’s USB port and using the free EasyLog software provided. Data can then be graphed, printed and exported to other applications for detailed analysis. Supplied with ½ AA battery and wall mount clip.