Learn More
Lascar Electronics Inc USB Vaccine Monitoring Kit
SDP
Catalog No. NC1629234
Encompass_Preferred
Lascar Electronics has paired its EL-USB-TP-LCD thermistor probe data logger with a glycol bottle thermistor probe for users who need a complete vaccine temperature monitoring system. The EL-USB-TP-LCD displays and records over 32,000 readings from the supplied thermistor probe. It features an LCD and push-button which allow the user to cycle through the most recent, highest and lowest stored temperature readings. EL-WIN-USB software enables quick configuration and data download via the USB port. Supplied with glycol bottle thermistor probe, long-life lithium battery, traceable calibration certificate and software.This product meets and exceeds CDC standards for vaccine monitoring.
Catalog No. NC1629234 Supplier Lascar Electronics Inc Supplier No. ELUSBVAC
Lascar Electronics has paired its EL-USB-TP-LCD thermistor probe data logger with a glycol bottle thermistor probe for users who need a complete vaccine temperature monitoring system. The EL-USB-TP-LCD displays and records over 32,000 readings from the supplied thermistor probe. It features an LCD and push-button which allow the user to cycle through the most recent, highest and lowest stored temperature readings. EL-WIN-USB software enables quick configuration and data download via the USB port. Supplied with glycol bottle thermistor probe, long-life lithium battery, traceable calibration certificate and software.This product meets and exceeds CDC standards for vaccine monitoring.