Learn More
Next Advance Inc Use FoamBlocker at 1 – 2% final concentration to reduce foaming during homogenization.
SDP
Catalog No. NC3913619
Encompass
FoamBlocker controls excess frothing or foaming of samples during homogenization
Catalog No. NC3913619 Supplier Next Advance Inc Supplier No. AFM2
FoamBlocker controls excess frothing or foaming of samples during homogenization