Learn More
Aviva Systems Biology Corp USE1 Antibody - middle region (ARP34134_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34134_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against USE1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human USE1. Alternate names: MDS032, SLT1, p31, P31ApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat, Zebrafish
Catalog No. 50-014-55263