Learn More
Bachem H-Arg(Pmc)-OtBu (free base)
SDP
Catalog No. 501947114
Encompass_Preferred
H-Arg(Pmc)-OtBu (free base), 169543-81-1, C24H40N4O5S, Mr 496.68, 5g. Used in the synthesis of peptide primers for cyanophycin, a cyanobacterial reserve polymer.
Catalog No. 50-194-7114 Supplier Bachem Supplier No. 4018591.0005
Add to Cart
Add to Cart
H-Arg(Pmc)-OtBu (free base), 169543-81-1, C24H40N4O5S, Mr 496.68, 5g. Used in the synthesis of peptide primers for cyanophycin, a cyanobacterial reserve polymer.