Learn More
New England Biolabs, Inc. USER® Enzyme – 50 units
SDP
Catalog No. 50811730
Encompass
USER (Uracil-Specific Excision Reagent) Enzyme generates a single nucleotide gap at the location of a uracil. USER Enzyme is a mixture of Uracil DNA glycosylase (UDG) and the DNA glycosylase-lyase Endonuclease VIII. UDG catalyses the excision of a uracil base, forming an abasic (apyrimidinic) site while leaving the phosphodiester backbone intact. The lyase activity of Endonuclease VIII breaks the phosphodiester backbone at the 3 and 5 sides of the abasic site so that base-free deoxyribose is released.Applications of USER include:
  • Directional RNA Seq
  • NEBNext adaptor cleavage
  • USER cloning
Catalog No. 50-811-730 Supplier New England Biolabs, Inc. Supplier No. M5505S
USER (Uracil-Specific Excision Reagent) Enzyme generates a single nucleotide gap at the location of a uracil. USER Enzyme is a mixture of Uracil DNA glycosylase (UDG) and the DNA glycosylase-lyase Endonuclease VIII. UDG catalyses the excision of a uracil base, forming an abasic (apyrimidinic) site while leaving the phosphodiester backbone intact. The lyase activity of Endonuclease VIII breaks the phosphodiester backbone at the 3 and 5 sides of the abasic site so that base-free deoxyribose is released.Applications of USER include:
  • Directional RNA Seq
  • NEBNext adaptor cleavage
  • USER cloning