Learn More
Aviva Systems Biology Corp NSD2 Antibody - N-terminal region (ARP34429_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34429_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against USF1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human USF1. Alternate names: UEF, FCHL, MLTF, FCHL1, MLTFI, HYPLIP1, bHLHb11, USF1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-55391