Learn More
Aviva Systems Biology Corp USF1 Antibody - middle region (ARP34707_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34707_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against USF1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human USF1. Alternate names: FCHL, FCHL1, HYPLIP1, MLTF, MLTFI, UEF, bHLHb11ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-55525