Learn More
Aviva Systems Biology Corp USF2 Antibody - N-terminal region (ARP33482_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33482_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against USF2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human USF2. Alternate names: FIP, bHLHb12ApplicationWB. Species reactivity: Cow, Guinea Pig, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-55041