Learn More
Ushio America, Inc. USHIO 14.7W MIDRANGE UV FL LMP
Catalog No. NC0931382
Encompass_Preferred
This item has a minimum qty of 3 per supplier requirements.
Uv-B (306Nm) Midrange Lamp. Watts - 14.7, :Ength - 436Mm, Diameter - 25.5Mm, Volts - 55V, Spectral Peak - 306, Uv Output - 3W, Base - G13, Average Life - 4000H.
Catalog No. NC0931382 Supplier Ushio America, Inc. Supplier No. 3000319
This item has a minimum qty of 3 per supplier requirements.
Uv-B (306Nm) Midrange Lamp. Watts - 14.7, :Ength - 436Mm, Diameter - 25.5Mm, Volts - 55V, Spectral Peak - 306, Uv Output - 3W, Base - G13, Average Life - 4000H.