Learn More
Bulbman Inc USHIO F4T5BL BLACKLIGHT 4W 6IN
SDP
Catalog No. 501253629
Encompass_Preferred
Ushio F4T5/BL: Ushio 4 watt 6" T5 Fluorescent bulb in black lite color has an opaque finish and features a miniature bi-pin base. Energy used: 4 W.Bulb shape: T5. Bulb base: Miniature bi-pin. - Average rated life: 3000 hrs. - Bulb finish: Opaque. - Produces UV-A light often used for nail polish curing, chemical processing, and insect trap applications. - Ushio 4 W 6" T5 Fluorescent bulb in black lite color has an average rated life of 3000 hrs and has a diameter of 0.625".
Catalog No. 50-125-3629 Supplier Bulbman Inc Supplier No. 113862
Ushio F4T5/BL: Ushio 4 watt 6" T5 Fluorescent bulb in black lite color has an opaque finish and features a miniature bi-pin base. Energy used: 4 W.Bulb shape: T5. Bulb base: Miniature bi-pin. - Average rated life: 3000 hrs. - Bulb finish: Opaque. - Produces UV-A light often used for nail polish curing, chemical processing, and insect trap applications. - Ushio 4 W 6" T5 Fluorescent bulb in black lite color has an average rated life of 3000 hrs and has a diameter of 0.625".