Learn More
Bulbman Inc Ushio F6T5/BL Black Light, 6 Watt
SDP
Catalog No. 501253632
Encompass_Preferred
Ushio F6T5/BL: Complement your interior with the Ushio 3000110 6-Watt, low-pressure, mercury-arc, black light lamp. This model is designed to emit radiation peaking at 352nm. It comes with a G5 base type and can be used for up to 3,000 hours. This Ushio lamp features low mercury and meets environmental demands.
Catalog No. 50-125-3632 Supplier Bulbman Inc Supplier No. 107662
Ushio F6T5/BL: Complement your interior with the Ushio 3000110 6-Watt, low-pressure, mercury-arc, black light lamp. This model is designed to emit radiation peaking at 352nm. It comes with a G5 base type and can be used for up to 3,000 hours. This Ushio lamp features low mercury and meets environmental demands.