Learn More
Bulbman Inc USHIO G20T10 GERMICIDAL BULB
SDP
Catalog No. 501253601
Encompass_Preferred
Ushio G20T10: Germicidal Lamp, T10, 20 watt, 58 volt, 8000 hour rated bulb with G13 base. Germicidal lamps are Low-Pressure Mercury-Arc lamps that emit radiation peaking at 253.7nm(UV-C). This output at 253.7nm is highly effective to inactive microorganisms such as algae, bacteria, virus, yeast and mold
Catalog No. 50-125-3601 Supplier Bulbman Inc Supplier No. 123601
Ushio G20T10: Germicidal Lamp, T10, 20 watt, 58 volt, 8000 hour rated bulb with G13 base. Germicidal lamps are Low-Pressure Mercury-Arc lamps that emit radiation peaking at 253.7nm(UV-C). This output at 253.7nm is highly effective to inactive microorganisms such as algae, bacteria, virus, yeast and mold