Learn More
Bulbman Inc Ushio G4T5, Germicidal Fluorescent Lamp Miniature Bi-Pin, Clear, 4 Watt
SDP
Catalog No. 501253585
Encompass_Preferred
Ushio G4T5: G4T5 - 4 WATT T5 GERMICIDAL BULB G5 BASE - 4 Watt T5 Clear Germicidal Fluorescent Lamp Miniature Bi-Pin (G5) Base 6.0" Maximum Overall Length, 6,000 Average Rated Hours. Spectral UV 253.7NM *These lamps are very dangerous, use only in fixtures which provide adequate protection. Avoid exposure. Ultraviolet radiation is harmful to the skin and the eyes and can cause serious skin burns and eye injury either from direct or reflected radiation. To reduce the risk of personal injury, install only in a fixture which provides adequate protection to area occupants. This bulb should not be used for illumination purposes. Do not operate this bulb without personal protective equipment, hand and eye protection should be worn at all times when handling.
Catalog No. 50-125-3585 Supplier Bulbman Inc Supplier No. 108270
Ushio G4T5: G4T5 - 4 WATT T5 GERMICIDAL BULB G5 BASE - 4 Watt T5 Clear Germicidal Fluorescent Lamp Miniature Bi-Pin (G5) Base 6.0" Maximum Overall Length, 6,000 Average Rated Hours. Spectral UV 253.7NM *These lamps are very dangerous, use only in fixtures which provide adequate protection. Avoid exposure. Ultraviolet radiation is harmful to the skin and the eyes and can cause serious skin burns and eye injury either from direct or reflected radiation. To reduce the risk of personal injury, install only in a fixture which provides adequate protection to area occupants. This bulb should not be used for illumination purposes. Do not operate this bulb without personal protective equipment, hand and eye protection should be worn at all times when handling.