Learn More
Bulbman Inc Ushio G6T5 Germicidal Fluorescent Lamp T5
SDP
Catalog No. 501253622
Encompass_Preferred
Ushio G6T5: USHIO Germicidal lamps are Low-Pressure Mercury-Arc lamps that emit radiation peaking at 253.7nm(UV-C). This output at 253.7nm is highly effective to inactivate microorganisms such as bacteria, virus, yeast and mold
Catalog No. 50-125-3622 Supplier Bulbman Inc Supplier No. 107647
Ushio G6T5: USHIO Germicidal lamps are Low-Pressure Mercury-Arc lamps that emit radiation peaking at 253.7nm(UV-C). This output at 253.7nm is highly effective to inactivate microorganisms such as bacteria, virus, yeast and mold