Learn More
Bulbman Inc Ushio G8T5E Germicidal UVB Linear Lamp T5
SDP
Catalog No. 501253564
Encompass_Preferred
Ushio G8T5E: The Ushio G8T5E is a a UVB linear lamp which emits ultraviolet rays between 280nm and 360nm (peak at 305nm-315nm), typically specified as 305nm, 306nm, 310nm, or 312nm.
Catalog No. 50-125-3564 Supplier Bulbman Inc Supplier No. 106149
Ushio G8T5E: The Ushio G8T5E is a a UVB linear lamp which emits ultraviolet rays between 280nm and 360nm (peak at 305nm-315nm), typically specified as 305nm, 306nm, 310nm, or 312nm.