Learn More
Bulbman Inc USHIO HBO100W/3
SDP
Catalog No. 501253545
Encompass_Preferred
Ushio HBO100W/3: HBO Mercury short-arc lamp without reflector. For any microscope model with HBO 100 lamp housing.
Catalog No. 50-125-3545 Supplier Bulbman Inc Supplier No. 131373
Ushio HBO100W/3: HBO Mercury short-arc lamp without reflector. For any microscope model with HBO 100 lamp housing.