Learn More
Medchemexpress LLC HY-114244 10mg Medchemexpress, USL311 CAS:1373268-67-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11424410MG
Encompass_Preferred
Medchemexpress, HY-114244 10mg USL311 CAS:1373268-67-7 USL311 is a selective CXCR4 antagonist, with anti-tumor activity. USL311 prevents the binding of stromal-cell derived factor-1 (SDF-1 or CXCL12) to CXCR4. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9311