Learn More
Cayman Chemical Usmarapride (oxalate), Purity: ≥98%, CAS: 1428862-33-2, 10 mg
SDP
Catalog No. 502910595
Encompass_Preferred
A 5-HT4 partial agonist; induces reporter gene expression in CHO cells expressing recombinant human 5-HT4E receptors (EC50 = 44 nM); increases novel object exploration time in rats at 1 and 3 mg/kg; reverses scopolamine-induced memory deficits in rats when administered at doses of 1, 3, and 10 mg/kg
Catalog No. 50-291-0595 Supplier Cayman Chemical Supplier No. 3586710 MG
A 5-HT4 partial agonist; induces reporter gene expression in CHO cells expressing recombinant human 5-HT4E receptors (EC50 = 44 nM); increases novel object exploration time in rats at 1 and 3 mg/kg; reverses scopolamine-induced memory deficits in rats when administered at doses of 1, 3, and 10 mg/kg