Learn More
Selleck Chemical LLC (+)-Usniacin 10mg 7562-61-0 D-Usnic Acid
SDP
Supplier:  Selleck Chemical LLC S225210MG
Encompass_Preferred
(+)-Usniacin (D-Usnic Acid) is a naturally occurring dibenzofuran derivative found in several lichen species. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-556-5