Learn More
Selleck Chemical LLC Usnic acid S4953-25mg
SDP
Catalog No. 503112668
Encompass_Preferred
Usnic acid (Usniacin) is a furandione found uniquely in lichen that is used widely in cosmetics deodorants toothpaste and medicinal creams as well as some herbal products It exhibits antiviral antiprotozoal antiproliferative anti-inflammatory and analgesic activity Usnic acid inhibits breast tumor angiogenesis and growth by suppressing VEGFR2-mediated AKT and ERK1/2 signaling pathways
Catalog No. 50-311-2668 Supplier Selleck Chemical LLC Supplier No. S495325MG
Add to Cart
Add to Cart
Usnic acid (Usniacin) is a furandione found uniquely in lichen that is used widely in cosmetics deodorants toothpaste and medicinal creams as well as some herbal products It exhibits antiviral antiprotozoal antiproliferative anti-inflammatory and analgesic activity Usnic acid inhibits breast tumor angiogenesis and growth by suppressing VEGFR2-mediated AKT and ERK1/2 signaling pathways