Learn More
Inorganic Ventures USP 232 ICH Q3D Class 1 Oral
SDP
Catalog No. NC1465333
Encompass_Preferred
USP<232>/ICH Q3D Class 1 Oral Elemental Impurities | Matrix: 10% v/v Nitric Acid | 125mL
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search
Catalog No. NC1465333 Supplier Inorganic Ventures Supplier No. IVSTOCK65125ML
USP<232>/ICH Q3D Class 1 Oral Elemental Impurities | Matrix: 10% v/v Nitric Acid | 125mL
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search