Learn More
U.S. Pharmacopeia Arginine Hydrochloride, 1119-34-2, MFCD00064550, 125mg
Supplier:  U.S. Pharmacopeia 1042601
Encompass
Molecular Formula C6H14N4O2.ClH, Molecular Weight 210.663, Melting Point 424.4 - 455 °F (218 - 235 °C) (decomposes), Boiling Point 766.4 - 770 °F (408 - 410 °C), Synonyms: Arginine monohydrochloride, L-(+)-Arginine monohydrochloride
Catalog No. NC0133336