Learn More
U.S. Pharmacopeia Dexamethasone, 50-02-2, MFCD00064136, 125mg
Catalog No. NC9196326
Encompass
Molecular formula C22H29FO5, Molecular weight 392.46, Melting Point 503.6 - 507.2 °F (262 - 264 °C)
Catalog No. NC9196326 Supplier U.S. Pharmacopeia Supplier No. 1176007
Add to Cart
Add to Cart
Molecular formula C22H29FO5, Molecular weight 392.46, Melting Point 503.6 - 507.2 °F (262 - 264 °C)