Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chem Service Inc USP Lanolin Pesticide Standard Mixture-1, 1 mL, 10 μg/mL, mixture in hexane, calibration mixture, 33 distinct analytes
SDP

Catalog No. NC2055818
Encompass_Preferred

This product is the USP Lanolin Pesticide Standard Mixture-1, provided by CHEM SERVICE INC, serving as a critical calibration mixture for various pesticide analyses. It is formulated as a mixture containing 33 distinct analytes, each at a concentration of 10 μg/mL, dissolved in Hexane. This standard is supplied in a 1 mL volume and comes with a Certificate of Analysis. For optimal stability, it should be stored at room temperature.

  • Product format is mixture
  • Concentration of 10 μg/mL
  • Solvent is hexane
  • Size is 1 mL
  • Intended use for calibration mixture
  • Product line is pesticide
  • Certificate of analysis included
  • Storage temperature range of 20 - 25 °C
  • Number of analytes is 33
  • Key analytes include 4,4'-DDD, 4,4'-DDE, 4,4'-DDT, Aldrin, Chlorpyrifos, Diazinon, Dieldrin, Endrin, Lindane

Catalog No. NC2055818 Supplier Chem Service Inc Supplier No. MUSPLAN1J101ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This product is the USP Lanolin Pesticide Standard Mixture-1, provided by CHEM SERVICE INC, serving as a critical calibration mixture for various pesticide analyses. It is formulated as a mixture containing 33 distinct analytes, each at a concentration of 10 μg/mL, dissolved in Hexane. This standard is supplied in a 1 mL volume and comes with a Certificate of Analysis. For optimal stability, it should be stored at room temperature.

  • Product format is mixture
  • Concentration of 10 μg/mL
  • Solvent is hexane
  • Size is 1 mL
  • Intended use for calibration mixture
  • Product line is pesticide
  • Certificate of analysis included
  • Storage temperature range of 20 - 25 °C
  • Number of analytes is 33
  • Key analytes include 4,4'-DDD, 4,4'-DDE, 4,4'-DDT, Aldrin, Chlorpyrifos, Diazinon, Dieldrin, Endrin, Lindane

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.