Learn More
U.S. Pharmacopeia Salicylic Acid, 69-72-7, MFCD00002439, 125mg
Catalog No. NC9996295
Encompass
Molecular formula C7H6O3, Molecular weight 138.12, Melting point 314.6 - 321.8 °F (157 - 161 °C) ; 76 ° C (sublimes), Boiling point 411.8 °F (211 °C) at 20 mm Hg, Flash point 315.00 °F (157.22 °C) Closed Cup
Catalog No. NC9996295 Supplier U.S. Pharmacopeia Supplier No. 1609002
Molecular formula C7H6O3, Molecular weight 138.12, Melting point 314.6 - 321.8 °F (157 - 161 °C) ; 76 ° C (sublimes), Boiling point 411.8 °F (211 °C) at 20 mm Hg, Flash point 315.00 °F (157.22 °C) Closed Cup