Learn More
Hach Company Hach USP System Suitability Kit (500 ppb), 3 x 40 mL VOA Bottles (Blank; 500 ppb Sucrose; 500 ppb Benzoquinone)
Catalog No. NC2044151 Shop All Hach Company Products
Encompass
The USP System Suitability Kit (500 ppb) provides ready-to-use standards for validating TOC and organic analysis systems. Supplied as three 40 mL VOA bottles - a blank, a sucrose standard, and a benzoquinone standard each at 500 ppb - the kit is designed for method validation and routine system suitability checks in groundwater, municipal water, and surface water testing.
  • Provides three prepared standards for system suitability: blank, sucrose, and benzoquinone at 500 ppb.
  • Supplied in 40 mL VOA bottles for TOC/organic analysis compatibility.
  • Enables method validation and routine system checks for water matrices.
  • Reduces preparation time and improves repeatability of validation runs.
  • Suitable for groundwater, municipal water, and surface water testing environments.
Catalog No. NC2044151 Supplier Hach Company Supplier No. 9459700
The USP System Suitability Kit (500 ppb) provides ready-to-use standards for validating TOC and organic analysis systems. Supplied as three 40 mL VOA bottles - a blank, a sucrose standard, and a benzoquinone standard each at 500 ppb - the kit is designed for method validation and routine system suitability checks in groundwater, municipal water, and surface water testing.
  • Provides three prepared standards for system suitability: blank, sucrose, and benzoquinone at 500 ppb.
  • Supplied in 40 mL VOA bottles for TOC/organic analysis compatibility.
  • Enables method validation and routine system checks for water matrices.
  • Reduces preparation time and improves repeatability of validation runs.
  • Suitable for groundwater, municipal water, and surface water testing environments.