Learn More
ERA WATERS CORP USP SYSTEM SUIT SET-500ML-HDPE
SDP
Catalog No. NC1862965
Encompass
"Each system suitability set contains (1) Reagent Water Rw, (1) 0.500 mg/L C from USP Sucrose Rs and (1) 0.500 mg/L C from USP 1,4-benzoquinone Rss.This CRM is produced under ISO 9001, ISO/IEC 17025 and ISO 17034 (previously Guide 34) accredited quality system "
Catalog No. NC1862965 Supplier ERA WATERS CORP Supplier No. 18140
"Each system suitability set contains (1) Reagent Water Rw, (1) 0.500 mg/L C from USP Sucrose Rs and (1) 0.500 mg/L C from USP 1,4-benzoquinone Rss.This CRM is produced under ISO 9001, ISO/IEC 17025 and ISO 17034 (previously Guide 34) accredited quality system "