Learn More
ERA WATERS CORP USP SYSTEM SUITABILITY SET
SDP
Catalog No. NC1862958
Encompass
"Each system suitability set contains (1) Reagent Water Rw, (1) 0.500 mg/L C from USP Sucrose Rs and (1) 0.500 mg/L C from USP 1,4-benzoquinone Rss.This CRM is produced under ISO 9001, ISO/IEC 17025 and ISO 17034 (previously Guide 34) accredited quality system."
Catalog No. NC1862958 Supplier ERA WATERS CORP Supplier No. 18000ERA
"Each system suitability set contains (1) Reagent Water Rw, (1) 0.500 mg/L C from USP Sucrose Rs and (1) 0.500 mg/L C from USP 1,4-benzoquinone Rss.This CRM is produced under ISO 9001, ISO/IEC 17025 and ISO 17034 (previously Guide 34) accredited quality system."