Learn More
ERA WATERS CORP USP SYSTEM SUITABILITY SET - H
SDP
Catalog No. NC2011931
Encompass
Set of three vials includes (1) Reagent Water Rw, (1) 0.500 mg/L C from USP Sucrose Rs and (1) 0.500 mg/L C from USP 1,4-benzoquinone Rss. acidified to pH <3 with HCl
Catalog No. NC2011931 Supplier ERA WATERS CORP Supplier No. 18004H
Set of three vials includes (1) Reagent Water Rw, (1) 0.500 mg/L C from USP Sucrose Rs and (1) 0.500 mg/L C from USP 1,4-benzoquinone Rss. acidified to pH <3 with HCl