Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ubiquitin-specific protease 1 (human), His-tagged | >95.0% | 100UG
SDP

Catalog No. 5000174456
Encompass_Preferred

Recombinant full-length human ubiquitin-specific protease 1 (USP1) expressed in E. coli and supplied as an N-terminal 6xHis-tagged lyophilized protein for research use. It is characterized by defined enzymatic activity, high purity, low endotoxin, and storage stability suitable for biochemical and cell biology assays.

  • Full-length human USP1 with N-terminal 6xHis tag.
  • Approximately 97 kDa molecular weight.
  • Specific activity >20 pmol/min/μg (Ub-AMC assay).
  • Purity >95% by reducing SDS-PAGE.
  • Endotoxin <1 EU/μg (LAL method).
  • Lyophilized formulation from PBS, pH 7.4.
  • Recommended storage at -20°C; reconstitute ≥100 μg/mL.

Catalog No. 50-001-74456 Supplier Medchemexpress LLC Supplier No. HYP702838100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant full-length human ubiquitin-specific protease 1 (USP1) expressed in E. coli and supplied as an N-terminal 6xHis-tagged lyophilized protein for research use. It is characterized by defined enzymatic activity, high purity, low endotoxin, and storage stability suitable for biochemical and cell biology assays.

  • Full-length human USP1 with N-terminal 6xHis tag.
  • Approximately 97 kDa molecular weight.
  • Specific activity >20 pmol/min/μg (Ub-AMC assay).
  • Purity >95% by reducing SDS-PAGE.
  • Endotoxin <1 EU/μg (LAL method).
  • Lyophilized formulation from PBS, pH 7.4.
  • Recommended storage at -20°C; reconstitute ≥100 μg/mL.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.