Learn More
Aviva Systems Biology Corp USP10 Antibody - middle region (ARP59302_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59302_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against USP10. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human USP10. Alternate names: KIAA0190, MGC2621, UBPOApplicationWB. Species reactivity: Cow, Human, Mouse, Rat
Catalog No. 50-286-653