Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ubiquitin carboxyl-terminal hydrolase 10 | >85.0% | 10UG
SDP

Catalog No. 5000177132
Encompass_Preferred

Recombinant human ubiquitin carboxyl-terminal hydrolase 10 (USP10) is a full-length deubiquitinating enzyme expressed in Sf9 insect cells and supplied with His and GST affinity tags. The protein is delivered as a filtered solution in HEPES/NaCl/glycerol/DTT buffer, intended for research use and stored frozen to preserve activity.

  • Full-length mature human protein (A2-L798).
  • Expressed in Sf9 insect cells for eukaryotic folding.
  • N-terminal His and GST tags for affinity purification.
  • Approximate molecular weight 114.7 kDa.
  • Purity >85.0% by reducing SDS-PAGE.
  • Supplied in 50 mM HEPES, 200 mM NaCl, 20% glycerol, 1 mM DTT, pH 7.5.
  • Store at -80°C and avoid repeated freeze-thaw cycles.

Catalog No. 50-001-77132 Supplier Medchemexpress LLC Supplier No. HYP70258410UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Recombinant human ubiquitin carboxyl-terminal hydrolase 10 (USP10) is a full-length deubiquitinating enzyme expressed in Sf9 insect cells and supplied with His and GST affinity tags. The protein is delivered as a filtered solution in HEPES/NaCl/glycerol/DTT buffer, intended for research use and stored frozen to preserve activity.

  • Full-length mature human protein (A2-L798).
  • Expressed in Sf9 insect cells for eukaryotic folding.
  • N-terminal His and GST tags for affinity purification.
  • Approximate molecular weight 114.7 kDa.
  • Purity >85.0% by reducing SDS-PAGE.
  • Supplied in 50 mM HEPES, 200 mM NaCl, 20% glycerol, 1 mM DTT, pH 7.5.
  • Store at -80°C and avoid repeated freeze-thaw cycles.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.