Learn More
Aviva Systems Biology Corp USP13 Antibody - N-terminal region (ARP59306_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59306_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against USP13. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human USP13. Alternate names: ISOT3, IsoT-3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-286-654