Learn More
Enzo Life Sciences USP14 (human), (recombinant) (untagged) (100 µg)
SDP
Catalog No. 501031802
Encompass_Preferred
Alternative name: Ubiquitin specific protease 14. MW: 56.1 kDa. Purity: ≥90% (SDS-PAGE). Formulation: Liquid. In 10mM TRIS containing 50mM NaCl and 1mM DTT, pH 7.4. Source: Produced in E. coli. UniProt: P54578. Long Term Storage: -80°C
Catalog No. 50-103-1802 Supplier Enzo Life Sciences Supplier No. BMLUW98400100
Alternative name: Ubiquitin specific protease 14. MW: 56.1 kDa. Purity: ≥90% (SDS-PAGE). Formulation: Liquid. In 10mM TRIS containing 50mM NaCl and 1mM DTT, pH 7.4. Source: Produced in E. coli. UniProt: P54578. Long Term Storage: -80°C