Learn More
LifeSensors USP15 25µg
SDP
Catalog No. 501530756
Encompass_Preferred
USP15 (Ubiquitin Specific Protease 15, UNPH4) is a deubiquitinase that associates with the COP9 signalosome (CSN) complex. It regulates various pathways such as the TGF-beta receptor signaling, NFκB and RNF41/NRDP1-PRKN pathways.
Catalog No. 50-153-0756 Supplier LifeSensors Supplier No. DB05130025
USP15 (Ubiquitin Specific Protease 15, UNPH4) is a deubiquitinase that associates with the COP9 signalosome (CSN) complex. It regulates various pathways such as the TGF-beta receptor signaling, NFκB and RNF41/NRDP1-PRKN pathways.