Learn More
Sino Biological Recombinant Human USP10 Protein (GST Tag), Active
Catalog No. 502572619
Encompass_Preferred
Recombinant human USP10 (154-798) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-257-2619 Supplier Sino Biological Supplier No. U510381G500
Recombinant human USP10 (154-798) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.