Learn More
Aviva Systems Biology Corp USP16 Antibody - N-terminal region (ARP45772_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45772_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against USP16. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human USP16. Alternate names: UBP-MApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-59134