Learn More
Aviva Systems Biology Corp USP17L2 Antibody - middle region (ARP59279_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59279_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against USP17L2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human USP17L2. Alternate names: DUB3, DUB-3ApplicationWB. Species reactivity: Human
Catalog No. 50-286-640