Learn More
Enzo Life Sciences USP19 polyclonal antibody (100 µl)
SDP
Supplier:  Enzo Life Sciences BMLPW10250100
Encompass_Preferred
Immunogen: Recombinant fragment of human USP19 (aa 1-129). Source: Purified from rabbit serum. Species reactivity: Human, Mouse, Rat. Applications: WB. Purity: Immunoaffinity purified. Formulation: Liquid. In 100mM TRIS, pH 7.0 containing 100mM glycine and 0.02% sodium azide.
Catalog No. 50-201-1562