Learn More
Enzo Life Sciences USP2 (catalytic domain) (rat), (recombinant) (untagged) (100 µg)
SDP
Catalog No. NC1347563
Encompass_Preferred
Alternative name: Ubiquitin specific protease 2. MW: ~40kDa. Purity: ≥95% (SDS-PAGE, following ion exchange chromatography). Formulation: Liquid. In 10mM MES, 0.5mM DTT, 50mM TRIS-HCl, pH 7.5, containing 20% glycerol. Source: Produced in E. coli BL21 (λDE3). Untagged USP2 catalytic domain. UniProt: Q5U349. Handling: Avoid freeze/thaw cycles. After opening, prepare aliquots and store at -80°C. Long Term Storage: -80°C
Catalog No. NC1347563 Supplier Enzo Life Sciences Supplier No. BMLUW98500100
Alternative name: Ubiquitin specific protease 2. MW: ~40kDa. Purity: ≥95% (SDS-PAGE, following ion exchange chromatography). Formulation: Liquid. In 10mM MES, 0.5mM DTT, 50mM TRIS-HCl, pH 7.5, containing 20% glycerol. Source: Produced in E. coli BL21 (λDE3). Untagged USP2 catalytic domain. UniProt: Q5U349. Handling: Avoid freeze/thaw cycles. After opening, prepare aliquots and store at -80°C. Long Term Storage: -80°C