Learn More
Aviva Systems Biology Corp USP22 Antibody - middle region (ARP58353_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58353_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against USP22. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human USP22. Alternate names: KIAA1063, USP3LApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53765