Learn More
Biomatik Corporation Usp22 Polyclonal Antibody
Supplier:  Biomatik Corporation CAC11158
Encompass_Preferred
Usp22 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to USP22. The Usp22 Polyclonal Antibody has been validated for the following applications: ELISA, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Ubiquitin Carboxyl-Terminal Hydrolase 22 Protein (330-478Aa). Applications: ELISA, IHC, IF. Target Synonyms: USP22. Accession Number: Q9UPT9. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4682