Learn More
TARGETMOL CHEMICALS INC USP22-IN-1 5MG
SDP
Catalog No. 502579220
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. USP22-IN-1 is a ubiquitin-specific peptidase 22 (USP22) inhibitor that can be used to treat proliferative diseases or cancer. purity: 98%
Catalog No. 50-257-9220 Supplier TARGETMOL CHEMICALS INC Supplier No. T731515MG
Add to Cart
Add to Cart
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. USP22-IN-1 is a ubiquitin-specific peptidase 22 (USP22) inhibitor that can be used to treat proliferative diseases or cancer. purity: 98%