Learn More
Selleck Chemical LLC Usp22i-S02-E1330-5MG
SDP
Supplier:  Selleck Chemical LLC E13305MG
Encompass_Preferred
Usp22i-S02(USP22-IN-1 compound S02) is a potent inhibitor of ubiquitin specific peptidase 22 (USP22) It showed anticancer activity by suppressing Foxp3 expression in T-regulatory cells
Catalog No. 50-238-9717