Learn More
Inorganic Ventures MUSP232 PRECIOUS MTLS STD 125m
SDP
Catalog No. 50272316
Encompass_Preferred
Inorganic Ventures' USP 232 Precious Metals Elemental Impurities is a 125mL certified reference material set in a hydrochloric acid matrix for stability. This standard is Traceable to NIST and manufactured in accordance with our stringent Quality Assurance guidelines. See Certificate of Analysis for expiration.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search
Catalog No. 50-272-316 Supplier Inorganic Ventures Supplier No. IVSTOCK38125ML
Inorganic Ventures' USP 232 Precious Metals Elemental Impurities is a 125mL certified reference material set in a hydrochloric acid matrix for stability. This standard is Traceable to NIST and manufactured in accordance with our stringent Quality Assurance guidelines. See Certificate of Analysis for expiration.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search