Learn More
Selleck Chemical LLC USP25/28 inhibitor AZ1-5mg
SDP
Catalog No. 502390042
Encompass_Preferred
AZ1 is a potent and noncompetitive dual Inhibitors of the ubiquitin-specific protease (USP) 25/28 with IC50s of 0.62 μM and 0.7 μM in Ub-RH110 assay,respectively.
Catalog No. 50-239-0042 Supplier Selleck Chemical LLC Supplier No. S89045MG
Add to Cart
Add to Cart
AZ1 is a potent and noncompetitive dual Inhibitors of the ubiquitin-specific protease (USP) 25/28 with IC50s of 0.62 μM and 0.7 μM in Ub-RH110 assay,respectively.