Learn More
Aviva Systems Biology Corp USP26 Antibody - middle region (ARP59318_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59318_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against USP26. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human USP26. Alternate names: MGC120066, MGC120067, MGC120068ApplicationWB. Species reactivity: Human
Catalog No. 50-286-659