Learn More
Aviva Systems Biology Corp Usp27x Antibody - middle region (ARP58354_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58354_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Usp27x. It was validated on Western Blot. . Alternate names: Sfc11, Usp27ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-286-465